Merck
  • One-pot synthesis of folic acid encapsulated upconversion nanoscale metal organic frameworks for targeting, imaging and pH responsive drug release.
One-pot synthesis of folic acid encapsulated upconversion nanoscale metal organic frameworks for targeting, imaging and pH responsive drug release.
Dalton transactions (Cambridge, England : 2003) (2016-10-28)
Angshuman Ray Chowdhuri, Dipranjan Laha, Srimanta Pal, Parimal Karmakar, Sumanta Kumar Sahu
ABSTRACT
In this work, a new theranostic nanoplatform is developed to construct an anticancer drug carrier by integrating the distinct advantages of upconversion nanoparticles (UCNPs) and metal organic frameworks (MOFs) encapsulated with a targeting ligand. Here NaYF
Sigma-Aldrich
Erbium(III) chloride, anhydrous, powder, 99.9% trace metals basis
Sigma-Aldrich
Ytterbium(III) chloride, anhydrous, powder, 99.9%
Sigma-Aldrich
Oleic acid, ≥99% (GC)