Merck
Prussian Blue Mg-Li Hybrid Batteries.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2016-11-08)
Xiaoqi Sun, Victor Duffort, Linda F Nazar
ABSTRACT
The major advantage of Mg batteries relies on their promise of employing an Mg metal negative electrode, which offers much higher energy density compared to graphitic carbon. However, the strong coulombic interaction of Mg
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis