Merck
  • Synthesis, Electrochemistry, and Photophysical Studies of Ruthenium(II) Polypyridine Complexes with D-π-A-π-D Type Ligands and Their Application Studies as Organic Memories.
Synthesis, Electrochemistry, and Photophysical Studies of Ruthenium(II) Polypyridine Complexes with D-π-A-π-D Type Ligands and Their Application Studies as Organic Memories.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-08-10)
Ming-Yi Leung, Sammual Yu-Lut Leung, Di Wu, Tao Yu, Vivian Wing-Wah Yam
ABSTRACT
A new class of ruthenium(II) polypyridine complexes with a series of D-π-A-π-D type (D=donor, A=acceptor) ligands was synthesized and characterized by
Sigma-Aldrich
Tetraphenylcyclopentadienone, 98%