Merck
  • Highly atom efficient guanylation of both aromatic and secondary amines catalyzed by simple lanthanide amides.
Highly atom efficient guanylation of both aromatic and secondary amines catalyzed by simple lanthanide amides.
The Journal of organic chemistry (2007-08-07)
Qinghai Li, Shaowu Wang, Shuangliu Zhou, Gaosheng Yang, Xiancui Zhu, Yuyu Liu
ABSTRACT
It is demonstrated that the cyclopentadienyl-free simple lanthanide amides [(Me(3)Si)(2)N](3)Ln(mu-Cl)Li(THF)(3)(Ln = La, Sm, Eu, Y, Yb) and Ln[N(SiMe(3))(2)]3 (Ln = Y, Yb) are highly efficient catalysts for the guanylation of both aromatic and secondary amines with a high activity under mild conditions. It is found that these catalysts are compatible with a wide range of solvents and substrates.
Sigma-Aldrich
N,N′-Di-tert-butylcarbodiimide, 99%