Merck
  • Peripubertal Stress With Social Support Promotes Resilience in the Face of Aging.
Peripubertal Stress With Social Support Promotes Resilience in the Face of Aging.
Endocrinology (2016-03-05)
Kathleen E Morrison, Sneha Narasimhan, Ethan Fein, Tracy L Bale
ABSTRACT
The peripubertal period of development is a sensitive window, during which adverse experiences can increase the risk for presentation of cognitive and affective dysfunction throughout the lifespan, especially in women. However, such experiences in the context of a supportive social environment can actually ameliorate this risk, suggesting that resilience can be programmed in early life. Affective disorders and cognitive deficits commonly emerge during aging, with many women reporting increased difficulty with prefrontal cortex (PFC)-dependent executive functions. We have developed a mouse model to examine the interaction between peripubertal experience and age-related changes in cognition and stress regulation. Female mice were exposed to peripubertal chronic stress, during which they were either individually housed or housed with social interaction. One year after this stress experience, mice were examined in tasks to access their cognitive ability and flexibility in stress reactive measures. In a test of spatial memory acquisition and reversal learning where aged females normally display a decreased performance, the females that had experienced stress with social interaction a year earlier showed improved performance in reversal learning, a measure of cognitive flexibility. Because peripuberty is a time of major PFC maturation, we performed transcriptomic and biochemical analysis of the aged PFC, in which long-term changes in microRNA expression and in myelin proteins were found. These data suggest that stress in the context of social support experienced over the pubertal window can promote epigenetic reprogramming in the brain to increase the resilience to age-related cognitive decline in females.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Anti-Somatostatin Receptor Type 1 Antibody, Chemicon®, from rabbit