Merck
  • Annulation of primary amines to piperazines and diazaspirocycles utilizing alpha-methyl benzyl resin.
Annulation of primary amines to piperazines and diazaspirocycles utilizing alpha-methyl benzyl resin.
Journal of combinatorial chemistry (2006-01-10)
Calum Macleod, Blanca I Martinez-Teipel, William M Barker, Roland E Dolle
ABSTRACT
The microwave-assisted solid-phase synthesis of piperazines, 3,9-diazaspiro[5.5]undecanes and 2,9-diazaspiro[5.5]undecanes is reported. The synthesis relies on the direct annulation of primary amines with resin-bound bismesylates. Critical to the success of this chemistry was the development of alpha-methyl benzyl carbamate resin linker. This resin permits the cleavage of the heterocycles under mildly acidic conditions, free of contaminating linker-derived N-alkylated byproducts.
Sigma-Aldrich
4-Amino-1-Boc-piperidine, 97%