Merck
  • Impact of Species and Variety on Concentrations of Minor Lipophilic Bioactive Compounds in Oils Recovered from Plum Kernels.
Impact of Species and Variety on Concentrations of Minor Lipophilic Bioactive Compounds in Oils Recovered from Plum Kernels.
Journal of agricultural and food chemistry (2016-01-06)
Paweł Górnaś, Magdalena Rudzińska, Marianna Raczyk, Inga Mišina, Arianne Soliven, Gunārs Lācis, Dalija Segliņa
ABSTRACT
The profile of bioactive compounds (carotenoids, tocopherols, tocotrienols, phytosterols, and squalene) in oils recovered from the kernels of 28 plum varieties of hexaploid species Prunus domestica L. and diploid plums Prunus cerasifera Ehrh. and their crossbreeds were studied. Oil yields in plum kernels of both P. cerasifera and P. domestica was in wide ranges of 22.6-53.1 and 24.2-46.9% (w/w) dw, respectively. The contents of total tocochromanols, carotenoids, phytosterols, and squalene was significantly affected by the variety and ranged between 70.7 and 208.7 mg/100 g of oil, between 0.41 and 3.07 mg/100 g of oil, between 297.2 and 1569.6 mg/100 g of oil, and between 25.7 and 80.4 mg/100 g of oil, respectively. Regardless of the cultivar, β-sitosterol and γ-tocopherol were the main minor lipophilic compounds in plum kernel oils and constituted between 208.5 and 1258.7 mg/100 g of oil and between 60.5 and 182.0 mg/100 g of oil, respectively. Between the studied plum species, significant differences were recorded for δ-tocopherol (p = 0.007), 24-methylenecycloartanol (p = 0.038), and citrostadienol (p = 0.003), but they were insufficient for discrimination by PCA.
Sigma-Aldrich
5-α-Cholestane, ≥97.0% (HPLC)
Sigma-Aldrich
Campesterol, ~65%