- Functional conservation between mammalian MGRN1 and plant LOG2 ubiquitin ligases.
Functional conservation between mammalian MGRN1 and plant LOG2 ubiquitin ligases.
FEBS letters (2013-09-17)
Damian D Guerra, Réjane Pratelli, Edward Kraft, Judy Callis, Guillaume Pilot
PMID24036454
ABSTRACT
Plant LOSS OF GDU 2 (LOG2) and Mammalian Mahogunin Ring Finger 1 (MGRN1) proteins are RING-type E3 ligases sharing similarity N-terminal to the RING domain. Deletion of this region disrupts the interaction of LOG2 with the plant membrane protein GLUTAMINE DUMPER1 (GDU1). Phylogenetic analysis identified two clades of LOG2/MGRN1-like proteins in vertebrates and plants. The ability of MGRN1 to functionally replace LOG2 was tested. MGRN1 ubiquitylates GDU1 in vitro and can partially substitute for LOG2 in the plant, partially restoring amino acid resistance to a GDU1-myc over-expression, log2-2 background. Altogether, these results suggest a conserved function for the N-terminal domain in evolution.