Merck
  • Expression of selected proteins of the extrinsic and intrinsic pathways of apoptosis in human leukocytes exposed to N-nitrosodimethylamine.
Expression of selected proteins of the extrinsic and intrinsic pathways of apoptosis in human leukocytes exposed to N-nitrosodimethylamine.
Human & experimental toxicology (2014-10-12)
A Iwaniuk, E Jabłońska, J Jabłoński, W Ratajczak-Wrona, M Garley
ABSTRACT
N-nitrosodimethylamine (NDMA) is a xenobiotic widespread in human environment capable of regulating the lifespan of immune cells. In this study, we examined the roles of the tumor necrosis factor-related apoptosis-inducing ligand (TRAIL)/death receptor 5 (DR5) complex and the Fas molecule in the induction of the extrinsic apoptosis pathway in human neutrophils (polymorphonuclear neutrophils (PMNs)) and peripheral blood mononuclear cells (PBMCs) exposed to NDMA. Also we assessed these proteins ability to trigger the intrinsic apoptosis pathway in those cells. For this purpose, we examined the expression of Fas-associated protein with death domain, truncated Bid (tBid) proteins, and apoptogenic factors such as apoptosis-inducing factor, Smac/Diablo, Omi/HtrA2, and caspase-3 as an indication of accomplished apoptosis phenomenon. PMNs and PBMCs were isolated from whole blood by density gradient centrifugation using Polymorphrep. Apoptotic cells were assessed with flow cytometry using a ready-made kit. The expression of proapoptotic molecules was investigated by Western blot analysis of PMNs and PBMCs treated with NDMA and/or rhTRAIL. The obtained results confirm the proapoptotic effects of NDMA on the examined human leukocytes and indicate an active participation of the TRAIL/DR5 complex and Fas protein in the process of apoptosis. Moreover, the research revealed distinct mechanisms of intrinsic apoptosis pathway activation between PMNs and PBMCs exposed to NDMA, as confirmed by the different levels of tBid, Smac/Diablo, Omi/HtrA2, and caspase-3 expression in those cells.