Merck
  • Periodontal status, inflammation, and malnutrition in hemodialysis patients - is there a link?
Periodontal status, inflammation, and malnutrition in hemodialysis patients - is there a link?
Journal of renal nutrition : the official journal of the Council on Renal Nutrition of the National Kidney Foundation (2014-09-15)
Liliana Garneata, Oana Slusanschi, Elena Preoteasa, Alexandra Corbu-Stancu, Gabriel Mircescu
ABSTRACT
Periodontal disease (PDD) was associated with inflammation, malnutrition, and higher mortality in hemodialysis (HD) patients. Cross-sectional observational study, aiming to assess the prevalence of PDD and the possible relationship among PDD, inflammation, and malnutrition in HD patients. Single HD center, 263 patients (age: 57.4 ± 12.3 years; 60% males; HD vintage 6.6 ± 4.9 years; the primary renal diseases were mainly primary glomerular nephropathies in 34% cases, with 11% diabetic nephropathy). Oral health status was assessed by the Silness and Loe plaque index, loss of clinical attachment level, periodontal pocket depth according to World Health Organization recommendations, by a single examiner. Patients were stratified by periodontal pocket depth (PPD): normal oral status/mild PDD (PPD < 4 mm), moderate PDD (PPD 4-5 mm), and severe PDD (PPD ≥ 6 mm). Demographic, smoking status, hematologic, dialysis-related data and parameters of the nutritional (Subjective Global Assessment score, anthropemetrical, and biochemical) and inflammatory status were collected. Poor periodontal status was shown by 75% of patients, 23% of them with severe PDD. Patients with PDD were older; higher percentages of them were smokers, diabetics, had malnutrition, and inflammation. Subjects with severe PDD had higher HD vintage, lower hemoglobin, and required higher darbepoetin doses than those with healthy periodontium. Darbepoetin resistance index was higher in patients with severe PDD than in those with normal periodontium. Models of multivariable linear logistic regression for the potential promoters and for the consequences of PDD revealed smoking and HD duration as significant contributors; increased C-reactive protein was associated with severe PDD. Cross-sectional observational design. Impaired periodontal health is highly prevalent in HD patients. PDD is more frequent in elderly diabetic smokers and in those with longer HD vintage; smoking and HD duration seems to be the most important determinants. The prevalence is higher in malnourished and in inflamed patients; inflammation seems to accompany PDD and to influence anemia response to treatment.