Merck
Design and implementation of a biomolecular concentration tracker.
ACS synthetic biology (2014-05-23)
Victoria Hsiao, Emmanuel L C de los Santos, Weston R Whitaker, John E Dueber, Richard M Murray
ABSTRACT
As a field, synthetic biology strives to engineer increasingly complex artificial systems in living cells. Active feedback in closed loop systems offers a dynamic and adaptive way to ensure constant relative activity independent of intrinsic and extrinsic noise. In this work, we use synthetic protein scaffolds as a modular and tunable mechanism for concentration tracking through negative feedback. Input to the circuit initiates scaffold production, leading to colocalization of a two-component system and resulting in the production of an inhibitory antiscaffold protein. Using a combination of modeling and experimental work, we show that the biomolecular concentration tracker circuit achieves dynamic protein concentration tracking in Escherichia coli and that steady state outputs can be tuned.
Sigma-Aldrich
Breathe-Easy® sealing membrane, polyurethane membrane with acrylic adhesive pre-cut to fit standard multiwell plates.