Merck
  • N-methylnicotinamide and nicotinamide N-methyltransferase are associated with microRNA-1291-altered pancreatic carcinoma cell metabolome and suppressed tumorigenesis.
N-methylnicotinamide and nicotinamide N-methyltransferase are associated with microRNA-1291-altered pancreatic carcinoma cell metabolome and suppressed tumorigenesis.
Carcinogenesis (2014-08-15)
Hui-Chang Bi, Yu-Zhuo Pan, Jing-Xin Qiu, Kristopher W Krausz, Fei Li, Caroline H Johnson, Chang-Tao Jiang, Frank J Gonzalez, Ai-Ming Yu
ABSTRACT
The cell metabolome comprises abundant information that may be predictive of cell functions in response to epigenetic or genetic changes at different stages of cell proliferation and metastasis. An unbiased ultra-performance liquid chromatography-mass spectrometry-based metabolomics study revealed a significantly altered metabolome for human pancreatic carcinoma PANC-1 cells with gain-of-function non-coding microRNA-1291 (miR-1291), which led to a lower migration and invasion capacity as well as suppressed tumorigenesis in a xenograft tumor mouse model. A number of metabolites, including N-methylnicotinamide, involved in nicotinamide metabolism, and l-carnitine, isobutyryl-carnitine and isovaleryl-carnitine, involved in fatty acid metabolism, were elevated in miR-1291-expressing PANC-1. Notably, N-methylnicotinamide was elevated to the greatest extent, and this was associated with a sharp increase in nicotinamide N-methyltransferase (NNMT) mRNA level in miR-1291-expressing PANC-1 cells. In addition, expression of NNMT mRNA was inversely correlated with pancreatic tumor size in the xenograft mouse model. These results indicate that miR-1291-altered PANC-1 cell function is associated with the increase in N-methylnicotinamide level and NNMT expression, and in turn NNMT may be indicative of the extent of pancreatic carcinogenesis.
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Chlorpropamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Supelco
Chlorpropamide, analytical standard, ≥97%
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water