Merck
  • Molecularly imprinted protein recognition thin films constructed by controlled/living radical polymerization.
Molecularly imprinted protein recognition thin films constructed by controlled/living radical polymerization.
Journal of bioscience and bioengineering (2014-07-26)
Shogo Sasaki, Tooru Ooya, Yukiya Kitayama, Toshifumi Takeuchi
ABSTRACT
We demonstrated the synthesis of molecularly imprinted polymers (MIPs) with binding affinity toward a target protein, ribonuclease A (RNase) by atom transfer radical polymerization (ATRP) of acrylic acid, acrylamide, and N,N'-methylenebisacrylamide in the presence of RNase. The binding activity of the MIPs was evaluated by surface plasmon resonance (SPR) of the MIP thin layers prepared on the gold-coated sensor chips. The MIPs prepared by ATRP (MIP-ATRP) had a binding affinity toward RNase with larger binding amount compared to MIPs prepared by conventional free radical polymerization methods (MIP-RP). Moreover, protein selectivity was evaluated using reference proteins (cytochrome c, myoglobin, and α-lactalbumin) and was confirmed in MIP-ATRP of optimum film thickness determined experimentally to be 15-30 nm; however, protein selectivity was not achieved in all MIP-RP. We have shown that ATRP is powerful technique for preparing protein recognition materials by molecular imprinting.
Sigma-Aldrich
11-Mercaptoundecanoic acid, 95%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
L-Ascorbic acid, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
2-(Dimethylamino)pyridine, 97%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
11-Mercaptoundecanoic acid, 98%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)