Merck
  • An expeditious entry to 9-azabicyclo[3.3.1]nonane N-oxyl (ABNO): another highly active organocatalyst for oxidation of alcohols.
An expeditious entry to 9-azabicyclo[3.3.1]nonane N-oxyl (ABNO): another highly active organocatalyst for oxidation of alcohols.
The Journal of organic chemistry (2009-05-30)
Masatoshi Shibuya, Masaki Tomizawa, Yusuke Sasano, Yoshiharu Iwabuchi
ABSTRACT
A practical, three-step synthetic route to 9-azabicyclo[3.3.1]nonane N-oxyl (ABNO, 3), an unhindered, stable class of nitroxyl radical, has been developed. ABNO exhibits a highly active nature compared with TEMPO in the catalytic oxidation of alcohols to their corresponding carbonyl compounds.
Sigma-Aldrich
9-Azabicyclo[3.3.1]nonane N-oxyl, 95%