Merck
  • Synthesis of cinnamyl ethers from α-vinylbenzyl alcohol using iodine as catalyst.
Synthesis of cinnamyl ethers from α-vinylbenzyl alcohol using iodine as catalyst.
Journal of oleo science (2010-09-30)
Yoshio Kasashima, Atsushi Uzawa, Kahoko Hashimoto, Tadasuke Nishida, Keiko Murakami, Takashi Mino, Masami Sakamoto, Tsutomu Fujita
ABSTRACT
Reactions of α-vinylbenzyl alcohol with other alcohols using iodine as a catalyst were investigated. The corresponding cinnamyl ethers were obtained as products. This suggested that α-vinylbenzyl alcohol was converted to cinnamyl ethers via 1-phenylallyl cation. Cinnamyl ethyl ether was obtained in 75% yield by the reaction of α-vinylbenzyl alcohol and ethanol in acetonitrile with iodine under the following conditions: temperature = 50 °C, molar ratio of α-vinylbenzyl alcohol:ethanol:iodine = 1:3.0:0.2, and time period = 6 h. Generally, the yields of the reactions using primary alcohols were higher than those using secondary and tertiary alcohols. Ether interchange also occurred by the reaction of α-vinylbenzyl alcohol and iodine, but proceeded smoothly only when an allyl group was used as the other substituent of the starting ether.
Sigma-Aldrich
α-Vinylbenzyl alcohol, 97%