Merck
  • Synthesis and characterization of transition metal complexes of 4-Amino-5-pyridyl-4H-1,2,4-triazole-3-thiol.
Synthesis and characterization of transition metal complexes of 4-Amino-5-pyridyl-4H-1,2,4-triazole-3-thiol.
SpringerPlus (2013-10-18)
Raghad Haddad, Emad Yousif, Ahmed Ahmed
ABSTRACT
Series of coordination complexes of Ni(II), Cu(II), Zn(II), Cd(II) and Sn(II) metal with 4-amino-5-(pyridyl)-4H-1,2,4-triazole-3-thiol, as a ligand has been successfully prepared in alcoholic medium. The prepared complexes were characterized quantitatively and qualitatively by using: microelemental analysis, FTIR spectroscopy, UV-visible spectroscopy, (1)H and (13)C NMR, magnetic susceptibility and conductivity measurements. This triazole ligand act as bidentate that coordination to the metal ions through sulphur and amine group. According to the spectral data of the complexes a tetrahedral geometry was suggested for these complexes except Cu (II) complexes which exhibit a square structure.
Sigma-Aldrich
4-Amino-5-(4-pyridyl)-4H-1,2,4-triazole-3-thiol, 97%