- CYB5R3: a key player in aerobic metabolism and aging?
CYB5R3: a key player in aerobic metabolism and aging?
Aging results from a complex and not completely understood chain of processes that are associated with various negative metabolic consequences and ultimately leads to senescence and death. The intracellular ratio of pyridine nucleotides (NAD(+)/NADH), has been proposed to be at the center stage of age-related biochemical changes in organisms, and may help to explain the observed influence of calorie restriction and energy-sensitive proteins on lifespan in model organisms. Indeed, the NAD(+)/NADH ratios affect the activity of a number of proteins, including sirtuins, which have gained prominence in the aging field as potential mediators of the beneficial effects of calorie restriction and mediating lifespan. Here we review the activities of a redox enzyme (NQR1 in yeast and CYB5R3 in mammals) that also influences the NAD(+)/NADH ratio and may play a regulatory role that connects aerobic metabolism with aging.