Merck
Cortactin signalling and dynamic actin networks.
The Biochemical journal (2004-06-10)
Roger J Daly
ABSTRACT
Cortactin was first identified over a decade ago, and its initial characterization as both an F-actin binding protein and v-Src substrate suggested that it was likely to be a key regulator of actin rearrangements in response to tyrosine kinase signalling. The recent discovery that cortactin binds and activates the actin related protein (Arp)2/3 complex, and thus regulates the formation of branched actin networks, together with the identification of multiple protein targets of the cortactin SH3 domain, have revealed diverse cellular roles for this protein. This article reviews current knowledge regarding the role of cortactin in signalling to the actin cytoskeleton in the context of these developments.
Sigma-Aldrich
Anti-Cortactin (p80/85) Antibody, clone 4F11, Alexa Fluor 488, clone 4F11, Upstate®, from mouse