Merck
  • Eosinophilic lung disease induced by bicalutamide: a case report and review of the medical literature.
Eosinophilic lung disease induced by bicalutamide: a case report and review of the medical literature.
Chest (1998-03-14)
P W Wong, N Macris, L DiFabrizio, N S Seriff
ABSTRACT
A 69-year-old man with advanced prostate cancer was receiving antiandrogen therapy (bicalutamide [Casodex]). He developed dyspnea, peripheral eosinophilia and bilateral pulmonary interstitial infiltrates. Transbronchial biopsy confirmed pulmonary eosinophilia. Withdrawal of bicalutamide and initiation of steroid therapy resulted in clinical improvement.
Sigma-Aldrich
Bicalutamide (CDX), ≥98% (HPLC), powder
Bicalutamide for system suitability, European Pharmacopoeia (EP) Reference Standard
Bicalutamide, European Pharmacopoeia (EP) Reference Standard