Merck
Gelatin-based delivery systems for cancer gene therapy.
Journal of drug targeting (2009-10-30)
Somayeh Hallaj Nezhadi, Peter F M Choong, Farzaneh Lotfipour, Crispin R Dass
ABSTRACT
Gelatin is a natural, biocompatible, nontoxic, edible, and inexpensive macromolecule. These properties result in its wide application in pharmaceutical, medical, and cosmetic products. Recently, it has been used for the delivery of such gene therapeutic entities as plasmid DNA. This review discusses the in vivo and in vitro studies using gelatin for delivery of therapeutic genes to cancerous cells. Recent studies show that present cancer gene therapy using gelatin is lacking in both efficiency and specificity in comparison with viral vectors, whereas complexes of therapeutic DNA with modified gelatin possibly offer a safe and efficient strategy for systemic administration of therapeutic genes to solid tumors compared to injection of naked plasmid DNA. The future of these promising approaches lies in the development of better techniques for preparing gelatin-gene complexes with the aim of a gelatin-based cancer gene therapy with comparable efficiency to viral vectors but with the added advantage of biosafety for patients.
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture