Merck
Recent developments in analytical determination of furosemide.
Journal of pharmaceutical and biomedical analysis (2008-08-15)
M Espinosa Bosch, A J Ruiz Sánchez, F Sánchez Rojas, C Bosch Ojeda
ABSTRACT
Furosemide (FUR), a drug that promotes urine excretion, is used in the pharmacotherapy of various diseases and is considered as a doping agent in sports. FUR is a powerful diuretic (water pill). This medicine is used to treat excessive fluid accumulation and swelling (edema) of the body caused by heart failure, cirrhosis, chronic kidney failure, and nephrotic syndrome. Owing to its extensive use as a powerful diuretic, FUR has long attracted the attention of many analysts. A variety of analytical methods have been proposed for the determination of FUR in biological fluids and pharmaceutical samples. The revision includes the most relevant analytical methodologies used in its determination from the nineties decade at present.
Product Description
Supelco
Furosemide, Pharmaceutical Secondary Standard; Certified Reference Material
Furosemide, European Pharmacopoeia (EP) Reference Standard
USP
Furosemide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Furosemide
Furosemide for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Furosemide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®