Merck
  • Photografting and patterning of oligonucleotides on benzophenone-modified boron-doped diamond.
Photografting and patterning of oligonucleotides on benzophenone-modified boron-doped diamond.
Chemical communications (Cambridge, England) (2007-07-05)
Sabine Szunerits, Naoto Shirahata, Paolo Actis, Jun Nakanishi, Rabah Boukherroub
ABSTRACT
Irradiation of a patterned benzophenone-terminated boron-doped diamond (BDD) surface with UV light (lambda = 350 nm) in the presence of a 15(mer) oligonucleotide resulted in the covalent linking of the DNA strand to the BDD interface.
Sigma-Aldrich
3-Benzoylbenzoic acid, 99%