Merck
  • Adverse events after unenhanced and monomeric and dimeric contrast-enhanced CT: a prospective randomized controlled trial.
Adverse events after unenhanced and monomeric and dimeric contrast-enhanced CT: a prospective randomized controlled trial.
Radiology (2006-05-25)
Hans H Schild, Christiane K Kuhl, Ute Hübner-Steiner, Ingrid Böhm, Ulrich Speck
ABSTRACT
To prospectively evaluate adverse events (AEs) in patients who underwent nonionic monomeric or dimeric contrast media-enhanced computed tomography (CT) and to compare these effects with AEs in patients who underwent unenhanced CT. Local ethics committee approval and written informed consent were obtained. Patients were randomly assigned to the dimeric group, monomeric group, or concurrent control group. Acute (occurring during or within 1 hour after contrast agent injection) and delayed (occurring between 1 hour and 7 days after contrast agent injection) AEs were evaluated and categorized (mild, not requiring treatment; moderate, self-limiting AE requiring simple treatment; severe, AE requiring extensive treatment or endangering life). The two-sided Fisher exact test and the Mann-Whitney U and Wilcoxon signed rank tests were used for statistical analysis. A total of 895 patients were recruited. Acute AEs were noted significantly (P < .05) more often in the monomeric group (44.8% [133 of 297 patients]) than in the dimeric (23.7% [71 of 300 patients]) or control (9.4% [28 of 298 patients]) groups. Two moderate acute AEs were noted in one patient in each contrast agent group; all other acute AEs were mild. There was no significant difference in the overall incidence of delayed AEs between the dimeric and monomeric groups (53.1% [139 of 262 patients] vs 50.8% [131 of 258 patients]). Delayed cutaneous AEs were noted significantly (P < .05) more often in the dimeric group. A total 16 AEs (2.0%) were moderate; no AE was severe. The dimeric contrast agent caused fewer acute AEs than the monomeric contrast agent; however, the dimeric and monomeric agents caused a similar overall number of delayed AEs. Delayed cutaneous symptoms were noted more often with the dimeric contrast agent. Both contrast agents were safe in that no severe AEs and only a few moderate AEs were observed.
Product Description
Iotrolan, European Pharmacopoeia (EP) Reference Standard
Iotrolan for system suitability, European Pharmacopoeia (EP) Reference Standard