Merck
  • New furofuran and butyrolactone lignans with antioxidant activity from the stem bark of Styrax japonica.
New furofuran and butyrolactone lignans with antioxidant activity from the stem bark of Styrax japonica.
Journal of natural products (2004-12-29)
Byung-Sun Min, Min-Kyun Na, Sei-Ryang Oh, Kyung-Seop Ahn, Gil-Saeng Jeong, Gao Li, Sang-Ku Lee, Hyouk Joung, Hyeong-Kyu Lee
ABSTRACT
A new furofuran lignan, styraxlignolide B (1), and four new dibenzyl-gamma-butyrolactone lignans, styraxlignolides C-F (2-5), were isolated from the EtOAc-soluble fraction of stem bark of Styrax japonica. Known compounds, taraxerol (6), syringin (7), and (-)-pinoresinol glucoside (8), were also obtained. The structures of styraxligonolides B-F were determined as 2alpha-(4'-hydroxy-3'-methoxyphenyl)-6alpha-(3' ',4' '-methylenedioxyphenyl)-8-oxo-3,7-dioxabicyclo[3.3.0]octane 4'-O-(beta-D-glucopyranoside) (1), (2S,3S)-2alpha-(3' '-hydroxy-4' '-methoxybenzyl)-3beta-(4'-hydroxy-3'-methoxybenzyl)-gamma-butyrolactone 4'-O-(beta-D-glucopyranoside) (2), (2S,3S)-2alpha-(4' '-hydroxy-3' '-methoxybenzyl)-3beta-(4'-hydroxy-3'-methoxybenzyl)-gamma-butyrolactone 4'-O-(beta-D-glucopyranoside) (3), (2S,3S)-2alpha-(4' '-hydroxy-3' '-methoxybenzyl)-3beta-(4'-hydroxy-3'-methoxybenzyl)-gamma-butyrolactone 4' '-O-(beta-D-glucopyranoside) (4), and (2S,3S)-2alpha-(3' ',4' '-dimethoxybenzyl)-3beta-(4'-hydroxy-3'-methoxybenzyl)-gamma-butyrolactone 4'-O-(beta-D-glucopyranoside) (5) by spectroscopic means including 2D NMR. Compounds 1-8 were tested in vitro for antioxidant activity against 1,1-diphenyl-2-picrylhydrazyl (DPPH) radicals. Styraxlignolide C (2), styraxlignolide D (3), styraxlignolide E (4), and (-)-pinoresinol glucoside (8) exhibited weak radical-scavenging activity in the DPPH assay, with IC50 values of 380, 278, 194, and 260 microM, respectively.
Product Description
Supelco
3β-Taraxerol, analytical standard