Merck
  • Association analysis of tapasin polymorphisms with aspirin-exacerbated respiratory disease in asthmatics.
Association analysis of tapasin polymorphisms with aspirin-exacerbated respiratory disease in asthmatics.
Pharmacogenetics and genomics (2013-06-06)
Sung-hwan Cho, Jong-Sook Park, Byung Lae Park, Da-Jeong Bae, Soo-Taek Uh, Mi-Kyeong Kim, Inseon S Choi, Hyoung Doo Shin, Choon-Sik Park
ABSTRACT
Aspirin-exacerbated respiratory disease (AERD) is characterized by the development of airway obstruction in asthmatic individuals following the ingestion of aspirin or other nonsteroidal anti-inflammatory drugs. TAPBP (TAP-binding protein, tapasin) is upregulated by eicosanoids, which act as potent inflammatory molecules in aspirin-related reactions. Thus, functional alterations in the TAPBP gene may contribute toward AERD. We examined the relationship between the single nucleotide polymorphisms on the TAPBP gene and AERD. A group of asthmatic patients (n=1252) underwent the oral aspirin challenge. Oral aspirin challenge reactions were categorized into two groups as follows: 15% or greater decreases in forced expiratory volume in 1 s or naso-ocular and skin reactions (AERD), or 15% or less decreases in forced expiratory volume in 1 s without naso-ocular and skin reactions (aspirin-tolerant asthma). Five single nucleotide polymorphisms of the TAPBP gene were genotyped. Logistic regression analysis showed that the minor allele frequencies of TAPBP rs2071888 C>G (Thr260Arg) on exon 4 (P>0.05), which was in absolute linkage disequilibrium with rs1059288 T>C on 3'UTR, were significantly higher in the AERD group than in the aspirin-tolerant asthma group, and the P values remained significant after multiple comparisons (Pcorr=0.006, odds ratio: 1.37, 95% confidence interval: 1.11-1.69, additive model; Pcorr=0.009, odds ratio: 1.52, 95% confidence interval: 1.14-2.03, dominant model). Alpha-helical wheel plotting showed that 260Arg had greater hydrophilic helical property than 260Thr. TAPBP polymorphisms may play a role in the development of AERD.
Sigma-Aldrich
Anti-TAPBP antibody produced in rabbit, affinity isolated antibody