Merck
Total synthesis of gigantetrocin A.
Chirality (2000-06-22)
Z M Wang, S K Tian, M Shi
ABSTRACT
A highly efficient synthetic method for the trans-tetrahydrofuran (THF) ring building block was established and the title compound was synthesized in 19 steps from trans-1,4-dichloro-2-butene via a convergent route with a Wittig reaction as the key step.
Sigma-Aldrich
trans-1,4-Dichloro-2-butene, technical grade, 85%, remainder predominantly cis isomer