Merck
  • Synthesis of a new class of carborane-containing star-shaped molecules via silicon tetrachloride promoted cyclotrimerization reactions.
Synthesis of a new class of carborane-containing star-shaped molecules via silicon tetrachloride promoted cyclotrimerization reactions.
Organic letters (2008-05-10)
Barada Prasanna Dash, Rashmirekha Satapathy, John A Maguire, Narayan S Hosmane
ABSTRACT
Symmetrical star-shaped molecules with carborane clusters on the periphery have been synthesized in good yields via silicon tetrachloride mediated cyclotrimerization reactions of 9-benzyl derivatives of carboranes with acetyl group substitution on the benzene ring. Facile functionalization of these symmetrical core structures with 1-iodoheptane and trivinylchlorosilane produce compounds which could be used as liquid crystalline substances and precursors for synthesis of higher order carbosilane dendrons.
Sigma-Aldrich
1-Iodoheptane, 98%