Merck
Surfactant ionic liquid-based microemulsions for polymerization.
Chemical communications (Cambridge, England) (2006-06-21)
Feng Yan, John Texter
ABSTRACT
Surfactants based on imidazolium ionic liquids (ILs), including polymerizable surfactant ILs, have been synthesized and used to stabilize polymerizable microemulsions useful for producing polymer nanoparticles, gels, and open-cell porous materials.
Sigma-Aldrich
11-Bromo-1-undecanol, 98%