Merck
  • Fully automated continuous flow synthesis of highly functionalized imidazo[1,2-a] heterocycles.
Fully automated continuous flow synthesis of highly functionalized imidazo[1,2-a] heterocycles.
Organic letters (2009-12-30)
Ananda Herath, Russell Dahl, Nicholas D P Cosford
ABSTRACT
The first continuous flow synthesis of imidazo[1,2-a]pyridine-2-carboxylic acids directly from 2-aminopyridines and bromopyruvic acid has been developed, representing a significant advance over the corresponding in-flask method. The process was applied to the multistep synthesis of imidazo[1,2-a]pyridine-2-carboxamides, including a Mur ligase inhibitor, using a two microreactor, multistep continuous flow process without isolation of intermediates.
Sigma-Aldrich
Bromopyruvic acid, ≥98.0%