Merck
  • Biological implications of the reaction possibilities of the proximate carcinogenic compound, N-hydroxy-2-fluorenylacetamide.
Biological implications of the reaction possibilities of the proximate carcinogenic compound, N-hydroxy-2-fluorenylacetamide.
Biomedical mass spectrometry (1976-02-01)
G Lhoëst, C Razzouk, M Mercier
ABSTRACT
The biological implications, the reaction possibilities and the structure of different derivatives of N-hydroxy-2-fluorenylacetamide have been described.
Product Description
Supelco
Trifluoroacetic anhydride, for GC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)