Merck
  • Novel chemoenzymatic strategy for the synthesis of enantiomerically pure secondary alcohols with sterically similar substituents.
Novel chemoenzymatic strategy for the synthesis of enantiomerically pure secondary alcohols with sterically similar substituents.
The Journal of organic chemistry (2003-06-21)
José-Luis Abad, Carles Soldevila, Francisco Camps, Pere Clapés
ABSTRACT
A novel chemoenzymatic strategy for the synthesis of enantiomerically pure secondary alcohols with sterically similar substituents is described. The key step is the kinetic lipase-catalyzed resolution of racemic mixtures of substituted propargylic alcohols. The efficiency of this new approach was tested in the preparation of the corresponding enantiomers of 1,11-hexadecandiol derivatives ((R)-5 and (S)-5). Two strategies were tested. In the first one, the racemic intermediate 1-octyn-3-ol (1) was resolved enzymatically and then elongated with 1-bromo-9,11-dioxadodecane. Alternatively, the racemic 1 can be elongated to the corresponding racemic 17,19-dioxa-7-eicosyn-6-ol (3) first and then resolved biocatalytically. Twelve commercially available lipases were screened for the kinetic resolution of these intermediates. Among them, Candida antarctica lipase (CAL-B) and Humicola lanuginosa lipase (HLL) were the best biocatalysts for the resolution of 1 (S enantiomer 90% ee, E = 35), and 3 (R enantiomer 90% ee, E = 34), respectively.
Sigma-Aldrich
1-Octyn-3-ol, 96%