Merck
The two faces of Cdk8, a positive/negative regulator of transcription.
Biochimie (2013-10-22)
Josipa Nemet, Branka Jelicic, Ivica Rubelj, Mary Sopta
ABSTRACT
Three cyclin dependent kinases, Cdk7, Cdk8 and Cdk9 are intimately connected with the processes of RNA polymerase II dependent transcription initiation and elongation in eukaryotic cells. Each of these kinases is part of a larger multisubunit complex, TFIIH, Mediator and p-TEFb respectively. Of the three kinases, Cdk8 is the most complex given that it has been associated with both positive and negative effects on transcription via mechanisms that include regulation of transcription factor turnover, regulation of CTD phosphorylation and regulation of activator or repressor function. Furthermore, Cdk8 has emerged as a key regulator of multiple transcriptional programs linked to nutrient/growth factor sensing and differentiation control. As such Cdk8 represents a potentially interesting therapeutic drug target. In this review we summarize the current state of knowledge on Cdk8 function both in yeast and higher eukaryotes as well as discussing the effects of Cdk8 null mutations at the organismal level.
Sigma-Aldrich
Anti-CDK8 antibody produced in rabbit, IgG fraction of antiserum