Merck
  • The Drosophila nuclear lamina protein YA binds to DNA and histone H2B with four domains.
The Drosophila nuclear lamina protein YA binds to DNA and histone H2B with four domains.
Molecular biology of the cell (2002-02-21)
Jing Yu, Mariana F Wolfner
ABSTRACT
Dramatic changes occur in nuclear organization and function during the critical developmental transition from meiosis to mitosis. The Drosophila nuclear lamina protein YA binds to chromatin and is uniquely required for this transition. In this study, we dissected YA's binding to chromatin. We found that YA can bind to chromatin directly and specifically. It binds to DNA but not RNA, with a preference for double-stranded DNA (linear or supercoiled) over single-stranded DNA. It also binds to histone H2B. YA's binding to DNA and histone H2B is mediated by four domains distributed along the length of the YA molecule. A model for YA function at the end of Drosophila female meiosis is proposed.
Sigma-Aldrich
Deoxyribonucleic acid, low molecular weight from salmon sperm