Merck
  • Endothelin activates large-conductance K+ channels in rat lactotrophs: reversal by long-term exposure to dopamine agonist.
Endothelin activates large-conductance K+ channels in rat lactotrophs: reversal by long-term exposure to dopamine agonist.
Endocrinology (1997-08-01)
B Kanyicska, M E Freeman, S E Dryer
ABSTRACT
Endothelin-1 (ET-1) inhibits PRL secretion from cultured rat lactotrophs. However, ET-1 stimulates PRL secretion after cultured lactotrophs have been exposed for 48 h to dopamine or D2 dopamine agonists. In the present study, we have used cell-attached and inside-out patch recordings to establish an ionic basis for these effects. Bath application of 20 nM ET-1 to untreated lactotrophs evoked a robust and persistent activation of large-conductance K+ channels in cell-attached patches. This effect of ET-1 had a long latency to onset, was maintained for as long as ET-1 was present, and required at least 10 min of washing in control saline before complete recovery was achieved. The stimulatory effect of 20 nM ET-1 on these channels was markedly attenuated in the presence of the selective ET(A) receptor antagonist BQ-610 (200 nM), or after pertussis toxin (200 ng/ml, 16 h) pretreatment. The unitary slope conductance of the ET-1 activated channels in cell attached patches was 165 and 95 pS when the recording electrodes contained 150 and 5.4 mM KCl, respectively. These channels were voltage-sensitive and their activity increased upon patch depolarization. Previously activated channels in cell-attached patches became quiescent immediately upon patch excision into Ca2+-free bath saline. Exposure of the intracellular surface to 0.1 microM Ca2+ restored the activity of these channels similar to the level seen before patch excision. In addition, preincubating the cells with the membrane-permeable Ca2+-chelator BAPTA-AM, or using Ca2+-free solution in the recording pipettes, prevented the activation of these channels by ET-1. The ET-1 activated large-conductance Ca2+-dependent K+ (BK(Ca)) channels were blocked by 20 mM tetraethylammonium but were insensitive to the K+ channel blockers apamin (1 microM), charybdotoxin (200 nM), or iberiotoxin (200 nM). Acute application of 10 microM dopamine and 20 nM ET-1 caused activation of BK(Ca) channels with indistinguishable kinetic properties, although the effect of dopamine occurred with shorter latency. After 48-h exposure to the specific D2 dopamine receptor agonist (+/-)-2-(N-phenyl-N-propyl) amino-5-hydroxytetralin hydrochloride (PPHT, 500 nM), bath application of 20 nM ET-1 resulted in inhibition of spontaneously active BK(Ca) channels. These data suggest that both the stimulatory and inhibitory effects of ET-1 on PRL secretion are mediated, at least in part, by actions on BK(Ca) channels, and that long term exposure to dopamine or D2 agonists alters the signaling pathways from the ET(A) receptor to BK(Ca) channels.