Merck
  • Fasting and decreased B cell sensitivity: important role for fatty acid-induced inhibition of PDH activity.
Fasting and decreased B cell sensitivity: important role for fatty acid-induced inhibition of PDH activity.
The American journal of physiology (1996-06-01)
Y P Zhou, D A Priestman, P J Randle, V E Grill
ABSTRACT
Fasting inhibits glucose-induced insulin secretion. We investigated the role of a glucose fatty acid cycle for such inhibition and its molecular basis in pancreatic islets from 48-h fasted rats. The fasting-impaired insulin response to 27 mM glucose was restored by 41% with a carnitine palmitoyltransferase I inhibitor, etomoxir. Etomoxir also restored (by 50%) impaired glucose oxidation in islets from fasted rats and increased the ratio of oxidation to glycolytic flux from 33 to 43%. Fasting decreased total pyruvate dehydrogenase (PDH) activity (active, unphosphorylated plus inactive, phosphorylated form) by 29%, as well as the percentage of active form (54 +/- 5 vs. 79 +/- 2% in fed rats, P < 0.001). Fasting increased islet PDH kinase activity as follows: PDH-bound activity by 36% and free (not PDH bound) PDH kinase by 70%. Fasting failed to affect PDH kinase content when assayed by an enzyme-linked immunoabsorbent assay with antibodies raised against 45 kDa PDH kinase alpha-chain. We conclude that fasting impairs B cell function to a major extent through the operation of a glucose fatty acid cycle and that decreased PDH activity resulting from increased specific activity of PDH kinase constitutes an important molecular mechanism.
Sigma-Aldrich
mono-Methyl hydrogen succinate, 95%