- [Thyroid function in patients with anorexia nervosa and depression].
[Thyroid function in patients with anorexia nervosa and depression].
Thyroid hormone levels were measured in 21 patients with anorexia nervosa, 15 patients with depression and 16 patients with severe depression and were compared with those in 53 normal subjects. In anorexia nervosa and severe depressed patients, serum T3, T4, fT3, fT4 and T3/T4 ratio showed significantly lower values than those in normal subjects. However there was no difference between depressed patients and normal subjects. The serum TSH levels were within normal range in all of the studied subjects. Thus, thyroid hormone levels in severe depressed patients were similar to those in anorexia nervosa and the changes were inversely related to disease conditions. The supplementation of thyroid hormones to antidepressant relieved clinical symptoms in some of the severe depressed patients. These results suggested that the changes in thyroid hormone levels in anorexia nervosa and severe depression were mainly due to impaired conversion of T4 to T3 by increased cortisol secretion through emotional stress.