- Effect of PGE2 and of agents that raise cAMP levels on macrophage activation induced by IFN-gamma and TNF-alpha.
Effect of PGE2 and of agents that raise cAMP levels on macrophage activation induced by IFN-gamma and TNF-alpha.
The effect of prostaglandin (PG) E2 on macrophage activation by interferon-gamma (IFN-gamma) and tumor necrosis factor-alpha (TNF-alpha) was evaluated. Murine macrophages infected with Leishmania enriettii or Leishmania major were activated by exposure to IFN-gamma (10-50 U/ml) and TNF-alpha (30-3000 U/ml), leading to intracellular parasite destruction within 24-48 h. Leishmanicidal activity was markedly increased when activation was performed in the presence of PGE2 (10(-9)-10(-7) M) or arachidonate (10(-5) M, a PG precursor), concomitant with enhanced nitrite release and glucose oxidation through the hexose monophosphate shunt pathway. Conversely, activation was reduced by indomethacin and hydrocortisone, two inhibitors of PG synthesis. Parasite killing and nitrite production were fully restored by exogenous PGE2, indicating that inhibition by these drugs was related to their ability to block PG production. PG can stimulate adenylate cyclase, thus raising intracellular cAMP levels. Accordingly, dibutyryl-cAMP, theophylline (which prevents cAMP breakdown), and forskolin (an activator of adenylate cyclase) all stimulated macrophage activation. Finally, PGE2 and cAMP enhanced expression of inducible nitric oxide synthase mRNA in response to IFN-gamma and TNF-alpha, and this effect was inhibited by the cAMP antagonist 2'-O-methyl adenosine. These findings are consistent with the hypothesis that PGE2 acts as a positive agonist in macrophage activation by IFN-gamma and TNF-alpha via its capacity to modulate intracellular cAMP levels.