Merck
  • Inhibition of arm regeneration by Ophioderma brevispina (Echinodermata, Ophiuroidea) by tributyltin oxide and triphenyltin oxide.
Inhibition of arm regeneration by Ophioderma brevispina (Echinodermata, Ophiuroidea) by tributyltin oxide and triphenyltin oxide.
Ecotoxicology and environmental safety (1986-08-01)
G E Walsh, L L McLaughlin, M K Louie, C H Deans, E M Lores
ABSTRACT
Effects of water-bourne toxicants on regeneration of arms by the brittle star, Ophioderma brevispina, are described. Regeneration was inhibited by 0.1 micrograms liter-1 bis(tri-n-butyltin)oxide and bis(triphenyltin)oxide. Both substances are known to act upon the nervous system, and it is suggested that inhibition was caused by neurotoxicological action of the tin compounds or by their direct effect upon tissue at the breakage point. The former is most likely because regeneration is mediated by the radial nerves of brittle stars.
Product Description
Supelco
Fentin hydroxide, PESTANAL®, analytical standard
Sigma-Aldrich
Triphenyltin hydroxide