- [Effect of the carcinogenic metabolites of aromatic amino acids on oxidative processes in lipids in vitro and in vivo].
[Effect of the carcinogenic metabolites of aromatic amino acids on oxidative processes in lipids in vitro and in vivo].
Biulleten' eksperimental'noi biologii i meditsiny (1987-07-01)
A A Levchuk, N P Pal'mina, M O Raushenbakh
PMID2441774
ABSTRACT
A comparative study of the effect produced by endogenous carcinogens (p-hydroxyphenyllactic and 5-methoxyindole-3-acetic acids) and their non-cancerogenic analogues on lipid peroxidation in vitro and in vivo was performed. It has been found that cancerogenic tyrosine and serotonin metabolites, unlike their non-cancerogenic analogues, increase lipid peroxidation in vitro. In vivo, cancerogenic tyrosine metabolite--p-hydroxyphenyllactic acid--is capable both of increasing and decreasing antioxidative lipid activity in the animal liver.
MATERIALS