Merck
  • Pollution by the fungicide pentachloronitrobenzene in an intensive farming area in Japan.
Pollution by the fungicide pentachloronitrobenzene in an intensive farming area in Japan.
The Science of the total environment (1990-03-01)
Y Fushiwaki, N Tase, A Saeki, K Urano
ABSTRACT
Environmental pollution by pentachloronitrobenzene (PCNB) was investigated at Tsumagoi, an intensive farming area, where a great amount of PCNB has been applied. High concentrations of PCNB were detected in river water near an area of cabbage cultivation. Further, pentachloroaniline (PCA) and pentachlorothioanisole (PCTA), which are the principal biodegradation products of PCNB, were also detected, and their ratios to PCNB were higher in autumn than in summer. PCNB concentrations in soil were similar to those of river sediment, in which PCNB was concentrated 5000-10,000 times over levels in river water. The biodegradation rate of PCNB in river water was higher than in river sediment and soil. The formation rates of PCA from PCNB were higher in river sediment and soil than in river water. It is considered that PCA remains for a long time in the environment and accumulates in river sediment and soil.
Product Description
Supelco
Pentachloroaniline, PESTANAL®, analytical standard