Merck
  • Optical phase conjugation by an As(2)S(3) glass planar waveguide for dispersion-free transmission of WDM-DPSK signals over fiber.
Optical phase conjugation by an As(2)S(3) glass planar waveguide for dispersion-free transmission of WDM-DPSK signals over fiber.
Optics express (2010-12-18)
M D Pelusi, F Luan, D-Y Choi, S J Madden, D A P Bulla, B Luther-Davies, B J Eggleton
ABSTRACT
We report the first demonstration of optical phase conjugation (OPC) transmission of phase encoded and wavelength-division multiplexed (WDM) signals by the Kerr effect in a planar structured waveguide. The phase conjugated electric field of the signal is produced by four wave mixing pumped by a CW laser during co-propagating with the signal in a highly nonlinear waveguide fabricated in As(2)S(3) glass. Experiments demonstrate the capability of the device to perform dispersion-free transmission through up to 225 km of standard single mode fiber for a 3 × 40 Gb/s WDM signal, with its channels encoded as return-to-zero differential phase shift keying and spaced either 100 or 200 GHz apart. This work represents an important milestone towards demonstrating advanced signal processing of high-speed and broadband optical signals in compact planar waveguides, with the potential for monolithic optical integration.