Merck
  • A sensitive LC-MS/MS method for the quantitative analysis of the Echinacea purpurea constituent undeca-2-ene-8,10-diynoic acid isobutylamide in human plasma.
A sensitive LC-MS/MS method for the quantitative analysis of the Echinacea purpurea constituent undeca-2-ene-8,10-diynoic acid isobutylamide in human plasma.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-12-03)
Andrew K L Goey, Rolf W Sparidans, Irma Meijerman, Hilde Rosing, Jan H M Schellens, Jos H Beijnen
ABSTRACT
Echinacea purpurea is one of the most popular herbal medicines and is known for its immunostimulatory effects. Alkylamides are the main lipophilic components of E. purpurea that contribute to its pharmacological actions. For quantification in human plasma of one of these alkylamides, undeca-2-ene-8,10-diynoic acid isobutylamide, a sensitive LC-MS/MS assay has been developed and validated. Plasma samples were pretreated using liquid-liquid extraction with a mixture of diethyl ether and n-hexane (50:50, v/v). Dried extracts were reconstituted in 50 μL of acetonitrile-water (50:50, v/v) after which 15 μL of sample was injected into the HPLC system. HPLC was performed using a Polaris 3 C18-A column (50 mm×2 mm ID) and isocratic elution with acetonitrile-water (50:50, v/v) containing 0.1% formic acid at a flow rate of 0.3 mL/min. Subsequently, electrospray ionization in the positive ion mode followed by tandem mass spectrometry was performed for detection. The total run time was 3 min. The assay was validated over a concentration range from 0.05 to 50 ng/mL for undeca-2-ene-8,10-diynoic acid isobutylamide, with 0.05 ng/mL being the lower limit of quantification using 1.0 mL plasma samples. Inter-assay inaccuracy (±12.7%), within-day and between-day precisions (CV≤8.23%) were acceptable. Further, undeca-2-ene-8,10-diynoic acid isobutylamide was found to be chemically stable under relevant conditions. Finally, the applicability of this assay has been successfully demonstrated in a pharmacokinetic experiment in which a human volunteer ingested a commercial extract of E. purpurea.
Sigma-Aldrich
Benzanilide, 98%