- Three factors that modulate the activity of class D β-lactamases and interfere with the post-translational carboxylation of Lys70.
Three factors that modulate the activity of class D β-lactamases and interfere with the post-translational carboxylation of Lys70.
The Biochemical journal (2010-11-27)
Lionel Vercheval, Cédric Bauvois, Alexandre di Paolo, Franck Borel, Jean-Luc Ferrer, Eric Sauvage, André Matagne, Jean-Marie Frère, Paulette Charlier, Moreno Galleni, Frédéric Kerff
PMID21108605
ABSTRACT
The activity of class D β-lactamases is dependent on Lys70 carboxylation in the active site. Structural, kinetic and affinity studies show that this post-translational modification can be affected by the presence of a poor substrate such as moxalactam but also by the V117T substitution. Val117 is a strictly conserved hydrophobic residue located in the active site. In addition, inhibition of class D β-lactamases by chloride ions is due to a competition between the side chain carboxylate of the modified Lys70 and chloride ions. Determination of the individual kinetic constants shows that the deacylation of the acyl-enzyme is the rate-limiting step for the wild-type OXA-10 β-lactamase.