Merck
  • An ATP-sensitive potassium channel blocker suppresses sodium-induced hypertension through increased secretion of urinary kallikrein.
An ATP-sensitive potassium channel blocker suppresses sodium-induced hypertension through increased secretion of urinary kallikrein.
Hypertension research : official journal of the Japanese Society of Hypertension (2009-03-06)
Yuji Kamata, Tomoe Fujita, Tetsuki Kato, Izumi Hayashi, Maya Kurosaka, Makoto Katori, Yoshikuni Fujita, Masataka Majima
ABSTRACT
It is suggested that an ATP-sensitive potassium channel blocker suppresses sodium-induced hypertension through increased secretion of urinary kallikrein. We reported that glibenclamide, an ATP-sensitive potassium channel blocker, accelerated dose-dependent secretion of renal kallikrein in sliced kidney cortex and in vivo in rats. In vehicle-treated normal Brown- Norway-Kitasato (nBN-Ki) rats, the administration of glibenclamide increased urinary kallikrein secretion, but changed neither the systolic blood pressure nor the urinary sodium on low (0.3%) NaCl diets. Although on high (8%) NaCl diets, the systolic blood pressure of the nBN-Ki rats administrated glibenclamide was significantly lower (P<0.05). The urinary levels of kallikrein and sodium of the nBN-Ki rats administrated glibenclamide were significantly increased (P<0.05, glibenclamide vs. vehicle). A similar result was obtained with a kidney-selective ATP-sensitive potassium blocker, N,N'-dicyclohexyl-4-morpholinecarboxamidine (U18177), in SD rats. Mutant kininogen-deficient Brown-Norway Katholiek (muBN-Ka) rats fed high (8%) NaCl diets showed an increase in urinary kallikrein levels, but showed neither hypotensive nor natriuretic actions by glibenclamide. A bradykinin B(2) receptor antagonist, 8-[3-[N-(E)-3-(6-acetamidopyridin-3-yl) acryloylglyycyl]-N-methylamino]-2,6-dichlorobenzyloxy]-2-methylquinoline (FR173657), which was administrated to SD rats, together with glibenclamide, abolished the hypotensive and natriuretic effects of glibenclamide in high-sodium (8%NaCl) hypertension, despite an accelerated secretion of urinary kallikrein. Therefore, these results indicate that glibenclamide, an ATP-sensitive potassium channel blocker suppressed sodium-induced hypertension through sodium excretion from the kidney resulting from accelerated secretion of urinary kallikrein.
Sigma-Aldrich
N,N′-Dicyclohexyl-4-morpholinecarboxamidine, 98%