Merck
  • Kinetics of blood flow after topical application of benzyl nicotinate on different anatomic sites.
Kinetics of blood flow after topical application of benzyl nicotinate on different anatomic sites.
Archives of dermatological research (2006-09-13)
Ute Jacobi, Marco Kaiser, Wolfram Sterry, Jürgen Lademann
ABSTRACT
Cutaneous characteristics, e.g., thickness of the SC and density of follicles, affect the penetration of topically applied substances. In the present study, the penetration of benzyl nicotinate, causing a vasodilation, was studied on three anatomic sites (forearm, forehead and calf) differing in their skin characteristics. Therefore, the blood flow of the superficial dermal plexus and that of the larger capillaries in the deeper skin layers was simultaneously measured using a laser Doppler flowmeter. In addition, the cutaneous temperature and redness were determined as a function of time. These four biological reactions were measured in turn on a skin area treated with a gel containing benzyl nicotinate and on an untreated control area. The highest basal levels were observed on the forehead. Topical application of benzyl nicotinate resulted in an increase in each biological response. Compared to the other sites, the maximal values were reached earliest on the forehead, which also showed the fastest decrease. No significant differences were obtained comparing the kinetic data of the calf with that of the forearm. The results indicate an additional contribution by the numerous vellus hair follicles of the forehead to the penetration and exposure of the drug.
Sigma-Aldrich
Benzyl nicotinate, ≥98.0% (GC)