Merck
  • Fatty acid esters of triterpenoids and steroid glycosides from Gambeya africana.
Fatty acid esters of triterpenoids and steroid glycosides from Gambeya africana.
Planta medica (2002-10-03)
Jean Wandji, Francois Tillequin, Dulcie A Mulholland, Jean-Duplex Wansi, Tanee Z Fomum, Victorine Fuendjiep, Francine Libot, Nole Tsabang
ABSTRACT
Chemical studies of the CH2 Cl2 -MeOH extract of the seeds of Gambeya africana (Baker) Pierre led to the isolation of 15 compounds. Their structures were established on the basis of spectroscopic methods and chemical reactions. They comprised five new fatty acid esters of erythrodiol, [3 beta-octacosanoyloxy-12-oleanen-28-ol, 3 beta-triacontanoyloxy-12-oleanen-28-ol, 3 beta-dotriacontanoyloxy-12-oleanen-28-ol, 3 beta-tetratriacontanoyloxy-12-oleanen-28-ol and 3 beta-hexatriacontanoyloxy-12-oleanen-28-ol], one new steroidal glycoside, [3 beta-O-beta-xylopyranosylchondrillasterol] and nine known compounds, (3 beta-octadecanoyloxy-12-oleanene, 3 beta-eicosanoyloxy-12-oleanene, 3 beta-docosanoyloxy-12-oleanene, 3 beta-acetoxy-12-oleanene, erythrodiol, 28-hydroxy-beta-amyrone, chondrillasterol, chondrillasterone and 3 beta-O-beta-glucopyranosylchondrillasterol).
Product Description
Supelco
Erythrodiol, analytical standard