Merck
Graphitic carbon in the Allende meteorite: a microstructural study.
Science (New York, N.Y.) (1981-04-17)
P P Smith, P R Buseck
ABSTRACT
High-resolution transmission electron microscopy, shows that carbon in the Allende carbonaceous chondrite meteorite is predominantly a poorly crystalline graphite. Such material is of interest as an important carrier of the isotopically anomalous noble gases found in carbonaceous chondrites.