Merck
  • Controlling the response of color tunable poly(N-isopropylacrylamide) microgel-based etalons with hysteresis.
Controlling the response of color tunable poly(N-isopropylacrylamide) microgel-based etalons with hysteresis.
Chemical communications (Cambridge, England) (2013-02-22)
Liang Hu, Michael J Serpe
ABSTRACT
We present the hysteresis of the response of the poly(N-isopropylacrylamide)-co-acrylic acid microgel-based etalons to solution temperature and pH changes. We find that the optical hysteresis of the etalon can be controlled using various solution ionic strengths and/or counterions, as well as by varying the microgel's acrylic acid concentration.
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Zinc acrylate, 98%