Merck
Inactivation of glutathione peroxidase by benzaldehyde.
Toxicology and applied pharmacology (1996-12-01)
T Tabatabaie, R A Floyd
ABSTRACT
Chronic benzaldehyde exposure is known to cause central nervous system (CNS) disturbances. Previous studies have shown that benzaldehyde causes the formation of reactive oxygen species (ROS) in rat synaptosomal fractions. Benzaldehyde has also been implicated in ROS formation in the CNS of rats treated with toluene. We have found that benzaldehyde effectively inactivates the antioxidant enzyme glutathione peroxidase (Ki approximately 15 microM), but has no effect on the other antioxidant enzymes tested: catalase, superoxide dismutase, and glutathione reductase. This effect has been found to be specific to benzaldehyde since other structurally related and unrelated aldehydes tested were found to be devoid of inactivating capacity toward glutathione peroxidase. Since glutathione peroxidase is the main enzyme responsible for removal of hydrogen peroxide and organic hydroperoxides in brain, its inactivation by benzaldehyde may be a main contributor to the observed ROS formation and the observed neurotoxicity caused by either benzaldehyde or toluene exposure.
Sigma-Aldrich
Selenocystamine dihydrochloride, powder