Merck
  • Effect of DI-n-pentyl phthalate treatment on testicular steroidogenic enzymes and cytochrome P-450 in the rat.
Effect of DI-n-pentyl phthalate treatment on testicular steroidogenic enzymes and cytochrome P-450 in the rat.
Toxicology letters (1983-02-01)
P M Foster, L V Thomas, M W Cook, D G Walters
ABSTRACT
Treatment of young male rats with dipentyl phthalate (DPP) produced significant decreases in testicular cytochrome P-450, cytochrome P-450 dependent microsomal steroidogenic enzymes (17 alpha-hydroxylase, 17-20 lyase) and in the maximal binding of a natural substrate (progesterone) to testis microsomes. No effect was demonstrated by this compound on hepatic cytochrome P-450 content. Treatment of animals with a phthalate ester not causing testicular atrophy (diethyl phthalate; DEP) produced no significant changes in any of the parameters measured. This effect on the enzymes responsible for androgen production may be important as a mechanism of action involved in the development of phthalate-induced testicular damage.
Product Description
Supelco
Dipentyl phthalate, for ion-selective electrodes, Selectophore, ≥99.0%